Learn More
Grainger Turnbuckle Body 800 lb Load Cap 2 1/4 L
Catalog No. 502768042
Encompass_Preferred
The Loos Locking Clip is a locking clip made of stainless steel with a width of 1/2 inch. It has a working load limit of 175 pounds and is used to secure pipes, cables, and hoses. The clip has a spring tension design that allows for tool-free installation and removal. It can accommodate items up to 1/2 inch in diameter. The locking clip meets military specifications and is made in the USA.
Catalog No. 50-276-8042 Supplier Grainger Supplier No. 16X749
The Loos Locking Clip is a locking clip made of stainless steel with a width of 1/2 inch. It has a working load limit of 175 pounds and is used to secure pipes, cables, and hoses. The clip has a spring tension design that allows for tool-free installation and removal. It can accommodate items up to 1/2 inch in diameter. The locking clip meets military specifications and is made in the USA.