Learn More
Grainger Kyocera Diamond Turning Insert, CBN, 80° Included Angle, 1/32 in. Corner Radius, 1 ea.
Catalog No. 502791308
Encompass_Preferred
These diamond indexable turning inserts, also known as rhombic inserts, are designed for general turning operations. Featuring two acute cutting points at opposite corners, they provide excellent access to complex contours and features. The inserts are intended for mounting to a compatible toolholder for use with a lathe or CNC machine and offer the ability to be indexed to utilize a fresh cutting edge.
  • Positive rake angle for efficient cutting
  • Diamond shape with an 80 ° included angle
  • Heat-resistant Megacoat finish
  • Constructed from CBN material for hardened steel applications
  • Suitable for boring, external turning, facing, and internal facing
  • Features a 7 ° clearance angle and 1/32 in. corner radius
  • Includes a chip-breaker for finishing applications
Catalog No. 50-279-1308 Supplier Grainger Supplier No. 167NL2
These diamond indexable turning inserts, also known as rhombic inserts, are designed for general turning operations. Featuring two acute cutting points at opposite corners, they provide excellent access to complex contours and features. The inserts are intended for mounting to a compatible toolholder for use with a lathe or CNC machine and offer the ability to be indexed to utilize a fresh cutting edge.
  • Positive rake angle for efficient cutting
  • Diamond shape with an 80 ° included angle
  • Heat-resistant Megacoat finish
  • Constructed from CBN material for hardened steel applications
  • Suitable for boring, external turning, facing, and internal facing
  • Features a 7 ° clearance angle and 1/32 in. corner radius
  • Includes a chip-breaker for finishing applications