Learn More
Grainger Turning Insert Diamond CNMG 433WE PV71
Catalog No. 502764162
Encompass_Preferred
The Grainger Approved CNMG 433WE-PV71 185GM8 is a turning insert made of carbide with a diamond shape. It has a CNMG style with a .433" IC size and is .185" thick. The grade is PV71 micrograin carbide with a PVD TiAlN coating. It is designed for light to medium turning of steels, stainless steels, cast iron, hardened steels, and exotic alloys. This insert features a positive 7 degree rake angle and is made in the USA.
Catalog No. 50-276-4162 Supplier Grainger Supplier No. 185GM8
The Grainger Approved CNMG 433WE-PV71 185GM8 is a turning insert made of carbide with a diamond shape. It has a CNMG style with a .433" IC size and is .185" thick. The grade is PV71 micrograin carbide with a PVD TiAlN coating. It is designed for light to medium turning of steels, stainless steels, cast iron, hardened steels, and exotic alloys. This insert features a positive 7 degree rake angle and is made in the USA.