Learn More
Grainger Turning Insert Diamond CNMG 432WF PV71
Catalog No. 502764648
Encompass_Preferred
The Grainger 185GM5 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings, manual reset thermal overload protection, and a Class B insulation system. The motor is CSA certified and can be used for a variety of light industrial applications. With its compact size and versatile performance, this Dayton motor is a reliable choice for many operations.
Catalog No. 50-276-4648 Supplier Grainger Supplier No. 185GM5
The Grainger 185GM5 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings, manual reset thermal overload protection, and a Class B insulation system. The motor is CSA certified and can be used for a variety of light industrial applications. With its compact size and versatile performance, this Dayton motor is a reliable choice for many operations.