Learn More
Grainger Kyocera Diamond Turning Insert, Carbide, 55° Angle, 10/Pk.
Catalog No. 502789758
Encompass_Preferred
These diamond indexable turning inserts, also known as rhombic inserts, are designed for general turning operations including back boring, copying, and external turning. They feature two acute cutting points at opposite corners that can be rotated to expose a fresh edge once the current one is worn. The pointed geometry facilitates access to complex contours and features that are difficult to machine with larger angle inserts.
  • Features positive rake with a 7° clearance angle
  • Designed with a medium finishing chip-breaker
  • Optimized for continuous to interrupted cutting applications
  • Constructed of carbide with a TiCN+Al2O3+TiN finish
  • Features excellent notching resistance and toughness
  • Primary application for stainless steel (ISO M) and secondary for high-temperature alloys (ISO S)
Catalog No. 50-278-9758 Supplier Grainger Supplier No. 167JV7
These diamond indexable turning inserts, also known as rhombic inserts, are designed for general turning operations including back boring, copying, and external turning. They feature two acute cutting points at opposite corners that can be rotated to expose a fresh edge once the current one is worn. The pointed geometry facilitates access to complex contours and features that are difficult to machine with larger angle inserts.
  • Features positive rake with a 7° clearance angle
  • Designed with a medium finishing chip-breaker
  • Optimized for continuous to interrupted cutting applications
  • Constructed of carbide with a TiCN+Al2O3+TiN finish
  • Features excellent notching resistance and toughness
  • Primary application for stainless steel (ISO M) and secondary for high-temperature alloys (ISO S)