Learn More
Grainger Kyocera Diamond Turning Insert, CBN, KBN05M, 1/64 in Radius, 1 ea
Catalog No. 502790191
Encompass_Preferred
This diamond turning insert is engineered for the precision machining of hardened steel. Utilizing a hybrid CBN grain structure that combines fine and coarse grains, it delivers superior hardness, strength, and thermal shock resistance. The insert features a specialized MEGACOAT finish and a molded chipbreaker design to effectively eliminate chip issues and enhance edge durability during demanding machining applications.
  • Molded chipbreaker design for excellent chip control in hardened materials
  • Hybrid CBN grain structure for improved wear and fracture resistance
  • Excellent sharpness with low-resistance edge preparation
  • MEGACOAT finish provides enhanced oxidation and wear resistance
  • Suitable for continuous to interrupted cutting applications
Catalog No. 50-279-0191 Supplier Grainger Supplier No. 167NM8
This diamond turning insert is engineered for the precision machining of hardened steel. Utilizing a hybrid CBN grain structure that combines fine and coarse grains, it delivers superior hardness, strength, and thermal shock resistance. The insert features a specialized MEGACOAT finish and a molded chipbreaker design to effectively eliminate chip issues and enhance edge durability during demanding machining applications.
  • Molded chipbreaker design for excellent chip control in hardened materials
  • Hybrid CBN grain structure for improved wear and fracture resistance
  • Excellent sharpness with low-resistance edge preparation
  • MEGACOAT finish provides enhanced oxidation and wear resistance
  • Suitable for continuous to interrupted cutting applications