Learn More
Grainger Turning Insert Diamond DNMX 431WF PV71
Catalog No. 502764183
Encompass_Preferred
The Sumitomo Diamond Turning Insert DNMX 52GP51 is a single-sided diamond turning insert for precision machining of non-ferrous metals and non-metallic materials. It features a 0.5mm nose radius and a 5 degree clearance angle. The insert is made of polycrystalline diamond brazed to a tungsten carbide substrate. It has an ISO designation of CNGA and is recommended for light to medium machining of aluminum, copper, graphite, plastics, and composites. The insert measures 7.938mm x 7.938mm x 3.175mm.
Catalog No. 50-276-4183 Supplier Grainger Supplier No. 185GM9
The Sumitomo Diamond Turning Insert DNMX 52GP51 is a single-sided diamond turning insert for precision machining of non-ferrous metals and non-metallic materials. It features a 0.5mm nose radius and a 5 degree clearance angle. The insert is made of polycrystalline diamond brazed to a tungsten carbide substrate. It has an ISO designation of CNGA and is recommended for light to medium machining of aluminum, copper, graphite, plastics, and composites. The insert measures 7.938mm x 7.938mm x 3.175mm.