Learn More
Grainger Turning Insert Diamond DNMX 443WF PV71
Catalog No. 502764340
Encompass_Preferred
The Grainger 4L185GN4 is an Allen Bradley 185GN4 Soft Starter, rated for 208-230V AC, 3 phase power. It has a current range of 14.4-29A and provides reduced voltage soft starting of 3 phase motors up to 3HP. This soft starter uses back-to-back SCRs to ramp up voltage and has adjustable ramp up time and initial torque. It has built-in overload protection and is designed for motor control applications where reduced starting current is necessary. The dimensions are 7.5 x 5.5 x 4.75 inches and it weighs approximately 5 pounds. This Allen Bradley soft starter is ideal for starting large electric motors smoothly.
Catalog No. 50-276-4340 Supplier Grainger Supplier No. 185GN4
The Grainger 4L185GN4 is an Allen Bradley 185GN4 Soft Starter, rated for 208-230V AC, 3 phase power. It has a current range of 14.4-29A and provides reduced voltage soft starting of 3 phase motors up to 3HP. This soft starter uses back-to-back SCRs to ramp up voltage and has adjustable ramp up time and initial torque. It has built-in overload protection and is designed for motor control applications where reduced starting current is necessary. The dimensions are 7.5 x 5.5 x 4.75 inches and it weighs approximately 5 pounds. This Allen Bradley soft starter is ideal for starting large electric motors smoothly.