Learn More
Grainger Turning Insert Diamond VBMT 222VF PV72
Catalog No. 502764287
Encompass_Preferred
The Walter Diamond Turning Insert VBMT 56TD53 is designed for precision machining of non-ferrous metals and non-metallic materials. It features a VBMT chipbreaker geometry, polycrystalline diamond composition, .250" IC size, .031" nose radius, 5° clearance angle, and is uncoated. Ideal for finishing cuts at low depths and feed rates up to .004 IPR, it uses a screw clamping system on CNC diamond turning lathes. It ensures excellent surface finish on materials like aluminum, copper, brass, plastics, and composites, allowing smooth, chatter-free machining.
Catalog No. 50-276-4287 Supplier Grainger Supplier No. 184ZT0
The Walter Diamond Turning Insert VBMT 56TD53 is designed for precision machining of non-ferrous metals and non-metallic materials. It features a VBMT chipbreaker geometry, polycrystalline diamond composition, .250" IC size, .031" nose radius, 5° clearance angle, and is uncoated. Ideal for finishing cuts at low depths and feed rates up to .004 IPR, it uses a screw clamping system on CNC diamond turning lathes. It ensures excellent surface finish on materials like aluminum, copper, brass, plastics, and composites, allowing smooth, chatter-free machining.