Learn More
Grainger Turning Insert Diamond VBMT 331VF PV72
Catalog No. 502764179
Encompass_Preferred
The Grainger 184ZT6 is a 6-inch diameter, 3-blade, zinc-plated steel propeller fan. It features a 1/3 HP, 115V, single-phase motor with a 1050 RPM operating speed and sleeve bearings. This fan produces airflow up to 1560 CFM and static pressure up to 0.25 inches. The fan housing is constructed of heavy-gauge steel with an enamel finish. This propeller fan is suitable for exhaust, cooling, circulation and ventilation applications.
Catalog No. 50-276-4179 Supplier Grainger Supplier No. 184ZT6
The Grainger 184ZT6 is a 6-inch diameter, 3-blade, zinc-plated steel propeller fan. It features a 1/3 HP, 115V, single-phase motor with a 1050 RPM operating speed and sleeve bearings. This fan produces airflow up to 1560 CFM and static pressure up to 0.25 inches. The fan housing is constructed of heavy-gauge steel with an enamel finish. This propeller fan is suitable for exhaust, cooling, circulation and ventilation applications.