Learn More
Grainger Turning Insert Diamond VBMT 333HQ PV72
Catalog No. 502764160
Encompass_Preferred
The WIDIA Diamond Turning Insert VBMT 433H27 is a diamond-tipped cutting tool designed for precision machining of non-ferrous metals and non-metallic materials. It features a CVD diamond cutting edge brazed to a carbide substrate. The insert has a .433" IC size, 55 degree diamond edge angle, and .250" max depth of cut. It is recommended for finishing cuts at low depths with speeds up to 2000 SFM and low to moderate feed rates. The diamond edge enables smooth, burr-free cutting of materials like aluminum, copper, graphite, and plastics. This single-sided insert with a .250" thickness is ideal for diamond turning applications.
Catalog No. 50-276-4160 Supplier Grainger Supplier No. 184ZU0
The WIDIA Diamond Turning Insert VBMT 433H27 is a diamond-tipped cutting tool designed for precision machining of non-ferrous metals and non-metallic materials. It features a CVD diamond cutting edge brazed to a carbide substrate. The insert has a .433" IC size, 55 degree diamond edge angle, and .250" max depth of cut. It is recommended for finishing cuts at low depths with speeds up to 2000 SFM and low to moderate feed rates. The diamond edge enables smooth, burr-free cutting of materials like aluminum, copper, graphite, and plastics. This single-sided insert with a .250" thickness is ideal for diamond turning applications.