Learn More
Grainger Turning Insert Diamond VBMT 3305VF PV7
Catalog No. 502764646
Encompass_Preferred
The Grainger 184ZT2 is a 2-ton capacity zinc-plated steel floor jack with a lifting range of 5-1/2 to 15 inches. It has a 45-degree tilt saddle to fit under low clearance vehicles and a universal joint release mechanism for precise load control. The floor jack has side handles and 4-1/2 inch front wheels for easy maneuverability. It meets or exceeds current ASME PALD standards.
Catalog No. 50-276-4646 Supplier Grainger Supplier No. 184ZT2
The Grainger 184ZT2 is a 2-ton capacity zinc-plated steel floor jack with a lifting range of 5-1/2 to 15 inches. It has a 45-degree tilt saddle to fit under low clearance vehicles and a universal joint release mechanism for precise load control. The floor jack has side handles and 4-1/2 inch front wheels for easy maneuverability. It meets or exceeds current ASME PALD standards.