Learn More
Grainger Turning Insert Diamond VBMT 332HQ PV72
Catalog No. 502764250
Encompass_Preferred
The KYOCERA Diamond Turning Insert TiCN 184ZT7 is a diamond turning insert made of titanium carbonitride (TiCN) with a grade of 184. It has a triangular shape with zero degrees of clearance angle and is 7mm thick. The insert features a CVD diamond coating and is designed for precision machining of non-ferrous metals like aluminum, copper, and their alloys. It provides a smooth mirror finish and can be used on CNC lathes at high cutting speeds with light depths of cut. The diamond coating reduces friction and enables a fine surface finish. This is a high performance insert for finish turning operations where an excellent surface finish is required.
Catalog No. 50-276-4250 Supplier Grainger Supplier No. 184ZT7
The KYOCERA Diamond Turning Insert TiCN 184ZT7 is a diamond turning insert made of titanium carbonitride (TiCN) with a grade of 184. It has a triangular shape with zero degrees of clearance angle and is 7mm thick. The insert features a CVD diamond coating and is designed for precision machining of non-ferrous metals like aluminum, copper, and their alloys. It provides a smooth mirror finish and can be used on CNC lathes at high cutting speeds with light depths of cut. The diamond coating reduces friction and enables a fine surface finish. This is a high performance insert for finish turning operations where an excellent surface finish is required.