Learn More
Grainger Turning Insert Diamond VBMT 332VF PV72
Catalog No. 502764251
Encompass_Preferred
The Grainger 184ZT9 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and sensitivity, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery life of up to 3000 hours. The Speedglas auto-darkening filter reacts in 1/25,000th of a second.
Catalog No. 50-276-4251 Supplier Grainger Supplier No. 184ZT9
The Grainger 184ZT9 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and sensitivity, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery life of up to 3000 hours. The Speedglas auto-darkening filter reacts in 1/25,000th of a second.