Learn More
Grainger Turning Insert Diamond VNMG 332VF PV72
Catalog No. 502764173
Encompass_Preferred
The Grainger 184ZV5 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This commercial fire extinguisher has a 2.5 lb capacity, is UL rated 2-B:C, and discharges in 10 seconds. It uses Novec 1230 fluid which leaves no residue and is ideal for protecting sensitive electronics. The extinguisher has a red steel cylinder with an enameled finish and comes equipped with a metal valve, pressure gauge, nylon strap, and steel handle. It measures approximately 5.4" W x 16.5" H. This clean agent fire extinguisher is well-suited for IT rooms, telecom spaces, industrial sites and vehicles.
Catalog No. 50-276-4173 Supplier Grainger Supplier No. 184ZV5
The Grainger 184ZV5 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This commercial fire extinguisher has a 2.5 lb capacity, is UL rated 2-B:C, and discharges in 10 seconds. It uses Novec 1230 fluid which leaves no residue and is ideal for protecting sensitive electronics. The extinguisher has a red steel cylinder with an enameled finish and comes equipped with a metal valve, pressure gauge, nylon strap, and steel handle. It measures approximately 5.4" W x 16.5" H. This clean agent fire extinguisher is well-suited for IT rooms, telecom spaces, industrial sites and vehicles.