Learn More
Grainger Kyocera Diamond Turning Insert, Carbide, Grade PR1225, 10/Pk.
Catalog No. 502791004
Encompass_Preferred
This carbide turning insert is designed for versatile machining applications including external turning, boring, internal facing, copying, and back boring. Engineered for high performance, it features a sharp edge with a polished finish and minus tolerance corner radius, providing three cutting edges for efficient material removal.
  • Features three cutting edges for extended tool life
  • Utilizes a superior wear and oxidation resistant MEGACOAT on a micro grain carbide substrate
  • Suitable for light interrupted to interrupted cutting of stainless steel
  • Designed with a sharp, polished edge for precision
Catalog No. 50-279-1004 Supplier Grainger Supplier No. 167NA2
This carbide turning insert is designed for versatile machining applications including external turning, boring, internal facing, copying, and back boring. Engineered for high performance, it features a sharp edge with a polished finish and minus tolerance corner radius, providing three cutting edges for efficient material removal.
  • Features three cutting edges for extended tool life
  • Utilizes a superior wear and oxidation resistant MEGACOAT on a micro grain carbide substrate
  • Suitable for light interrupted to interrupted cutting of stainless steel
  • Designed with a sharp, polished edge for precision