Learn More
Grainger Turning Insert Trigon WNMG 431WF PV710
Catalog No. 502764417
Encompass_Preferred
The Grainger 185GN8 is a 1/4 HP, 115V, 60Hz, 1725 RPM, single phase, open drip proof fan motor with a 48 frame size. It has a sleeve bearing and is thermally protected. The motor is totally enclosed and fan cooled (TEFC) with a cast iron body. It has a 1/2" diameter shaft and measures approximately 5-3/4" long by 5-1/4" wide by 6-1/2" high. The operating amps are 3.6 at full load. This electric motor is designed for continuous duty applications.
Catalog No. 50-276-4417 Supplier Grainger Supplier No. 185GN8
The Grainger 185GN8 is a 1/4 HP, 115V, 60Hz, 1725 RPM, single phase, open drip proof fan motor with a 48 frame size. It has a sleeve bearing and is thermally protected. The motor is totally enclosed and fan cooled (TEFC) with a cast iron body. It has a 1/2" diameter shaft and measures approximately 5-3/4" long by 5-1/4" wide by 6-1/2" high. The operating amps are 3.6 at full load. This electric motor is designed for continuous duty applications.