Learn More
Grainger Turning Insert Trigon WNMG 432WE PV710
Catalog No. 502764434
Encompass_Preferred
The Grainger 185GP1 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase open drip proof motor with a 48Y frame and features sleeve bearings for quiet operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected. The Dayton brand offers a 1 year warranty on this motor. With a standard NEMA 56C face and 1/2" diameter shaft, this electric motor can be used for a variety of light industrial, commercial, or residential applications.
Catalog No. 50-276-4434 Supplier Grainger Supplier No. 185GP1
The Grainger 185GP1 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase open drip proof motor with a 48Y frame and features sleeve bearings for quiet operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected. The Dayton brand offers a 1 year warranty on this motor. With a standard NEMA 56C face and 1/2" diameter shaft, this electric motor can be used for a variety of light industrial, commercial, or residential applications.