Learn More
Grainger Turning Insert Trigon WNMG 432WE PV720
Catalog No. 502764161
Encompass_Preferred
The Grainger 185GM2 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase open drip proof motor with a 48Y frame and features permanent split capacitor for high starting torque. The motor has ball bearings and a thermally protected design. It can be used for driving pumps, fans, blowers, and other shop tools and machinery. The motor dimensions are 6-1/2" diameter x 8-3/8" long and it weighs 15 pounds.
Catalog No. 50-276-4161 Supplier Grainger Supplier No. 185GM2
The Grainger 185GM2 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase open drip proof motor with a 48Y frame and features permanent split capacitor for high starting torque. The motor has ball bearings and a thermally protected design. It can be used for driving pumps, fans, blowers, and other shop tools and machinery. The motor dimensions are 6-1/2" diameter x 8-3/8" long and it weighs 15 pounds.