Learn More
Grainger Turning Insert Trigon WNMG 432WF PV710
Catalog No. 502764649
Encompass_Preferred
The Grainger 185GN9 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings for maintenance-free operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected against overloads. With its standard NEMA mounting, this general purpose motor is suitable for a variety of industrial, commercial and residential applications.
Catalog No. 50-276-4649 Supplier Grainger Supplier No. 185GN9
The Grainger 185GN9 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings for maintenance-free operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected against overloads. With its standard NEMA mounting, this general purpose motor is suitable for a variety of industrial, commercial and residential applications.