Learn More
TARGETMOL CHEMICALS INC TUROFEXORATE ISOPROPYL 25MG
SDP
Catalog No. 502579610
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Turofexorate Isopropyl (XL335) is a potent selective and orally bioavailable FXR agonist with EC50 of 4 nMpurity: 99%
Catalog No. 50-257-9610 Supplier TARGETMOL CHEMICALS INC Supplier No. T605325MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Turofexorate Isopropyl (XL335) is a potent selective and orally bioavailable FXR agonist with EC50 of 4 nMpurity: 99%