Learn More
Medchemexpress LLC HY-50911 5mg Medchemexpress, Turofexorate isopropyl CAS:629664-81-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY509115MG
Encompass_Preferred
Medchemexpress, HY-50911 5mg Turofexorate isopropyl CAS:629664-81-9 Turofexorate isopropyl (FXR-450) is a potent, selective, and orally bioavailable FXR agonist with EC50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2259