Learn More
Medchemexpress LLC Turofexorate isopropyl | 629664-81-9 | 99.8% | C25H24F2N2O3 | 1 ML
SDP
Catalog No. 5000357718
Encompass_Preferred
Turofexorate isopropyl (FXR-450) is a potent, selective, and orally bioavailable FXR agonist with an EC50 of 4 nM. It is intended for research and analytical applications, and an analytical standard is also available.
  • Potent, selective, and orally bioavailable FXR agonist
  • Used in autophagy, metabolic disease, and cancer research
  • Consistent with structure via 1H NMR and MS
  • High purity of 99.8%
Catalog No. 50-003-57718 Supplier Medchemexpress LLC Supplier No. HY5091110MM/1ML
Turofexorate isopropyl (FXR-450) is a potent, selective, and orally bioavailable FXR agonist with an EC50 of 4 nM. It is intended for research and analytical applications, and an analytical standard is also available.
  • Potent, selective, and orally bioavailable FXR agonist
  • Used in autophagy, metabolic disease, and cancer research
  • Consistent with structure via 1H NMR and MS
  • High purity of 99.8%