Learn More
Medchemexpress LLC Turofexorate isopropyl | 629664-81-9 | 99.8% | C25H24F2N2O3 | 25 MG
SDP
Catalog No. 5000358070
Encompass_Preferred
Turofexorate isopropyl is a highly potent, selective, and orally bioavailable agonist of the Farnesoid X Receptor (FXR).
  • Acts as an agonist for the Farnesoid X Receptor (FXR).
  • Exhibits significant potency with an EC50 of 4 nM.
  • Characterized by its selective action on FXR.
  • Orally bioavailable, making it suitable for certain research applications.
  • Utilized in research contexts related to Autophagy, Metabolic Disease, and Cancer.
  • Primarily intended for research and analytical applications.
Catalog No. 50-003-58070 Supplier Medchemexpress LLC Supplier No. 25MGHY50911
Turofexorate isopropyl is a highly potent, selective, and orally bioavailable agonist of the Farnesoid X Receptor (FXR).
  • Acts as an agonist for the Farnesoid X Receptor (FXR).
  • Exhibits significant potency with an EC50 of 4 nM.
  • Characterized by its selective action on FXR.
  • Orally bioavailable, making it suitable for certain research applications.
  • Utilized in research contexts related to Autophagy, Metabolic Disease, and Cancer.
  • Primarily intended for research and analytical applications.