Learn More
Medchemexpress LLC Turofexorate isopropyl | 629664-81-9 | 99.8% | C25H24F2N2O3 | 50 MG
SDP
Catalog No. 5000358363
Encompass_Preferred
Turofexorate isopropyl, also known as FXR-450, is a potent, selective, and orally bioavailable FXR agonist. It exhibits an EC50 of 4 nM and is stable under recommended storage conditions. This product is suitable for various laboratory and research applications, particularly in studies related to metabolic disease and cancer.
  • Potent, selective, and orally bioavailable FXR agonist
  • Features an EC50 of 4 nM
  • Stable under recommended storage conditions
  • High purity of 99.77% as determined by HPLC
  • Consistent with structure based on ¹H NMR spectrum and MS data
  • Applicable in metabolic disease and cancer research
Catalog No. 50-003-58363 Supplier Medchemexpress LLC Supplier No. HY5091150MG
Turofexorate isopropyl, also known as FXR-450, is a potent, selective, and orally bioavailable FXR agonist. It exhibits an EC50 of 4 nM and is stable under recommended storage conditions. This product is suitable for various laboratory and research applications, particularly in studies related to metabolic disease and cancer.
  • Potent, selective, and orally bioavailable FXR agonist
  • Features an EC50 of 4 nM
  • Stable under recommended storage conditions
  • High purity of 99.77% as determined by HPLC
  • Consistent with structure based on ¹H NMR spectrum and MS data
  • Applicable in metabolic disease and cancer research