Learn More
Aviva Systems Biology Corp TUT1 Antibody - N-terminal region (ARP33292_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TUT1. Alternate names: FLJ21850, FLJ22267, FLJ22347, MGC131987, MGC149809, PAPD2, RBM21, TUTase, STARPAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-54948