Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Tuttnauer™ Lab Line Benchtop Autoclave
SDP

Catalog No. 13890051
Click to view available options
Capacity (Metric):
28 L
65 L
85 L
160 L
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Benchtop autoclave designed to sterilize glassware, plastics, liquids and media.

Standard Operating Features

  • Designed to provide high quality, repeatable performance and accountability for a wide range of applications used in modern laboratories
  • Ideal for laboratory applications in research institutes, universities, pharmaceutical, biotechnology, food, and chemical industries
  • Benchtop lab autoclaves save space on crowded laboratory workbenches
  • For use with Agar preparation as well as the sterilization of liquids, pipettes, glass, instruments, and biohazards with various cooling options
  • Two integrated temperature probes for complete sterilization of liquids
  • Four preprogrammed cycles – glass, plastic, liquid A, and liquid B
  • Create up to 16 custom cycles and store up to 200 past cycles that you can open on your PC
  • Temperature selection from 105°C - 137°C (221°F - 279°F)
  • Water Saving System recycles used water after every cycle
  • Plug-n-play design: just add water to the reservoir and plug it in
  • Does not require connection to building utilities
  • PID (Proportional Integral Differential) pressure control
  • Built-in memory stores the previous 200 cycles
  • Multi-color display for easier reading, indicates stage of the cycle
  • Quick access to important information
  • Purchase includes autoclave stand, trays and printer

Service Plan

Extended warranty available

Warranty and Services

3-year warranty

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance UL, cUL, ASME
Capacity (English) 17 gal.
Capacity (Metric) 65 L
Depth (Metric) Chamber 45 cm
Dimensions (L x W) 15 x 19.7 in. (Chamber)
Diameter (English) Chamber 11 in.
Depth (English) Chamber 27.6 in.
Diameter (Metric) Chamber 28 cm
For Use With (Application) Sterilization of liquids, media, instruments, glassware
For Use With (Equipment) Liquids, Glassware, Utensils (Wrapped or Unwrapped), and Solids
Length (English) 30 in.
Height (English) Exterior 17.3 in.
Height (Metric) Exterior 44 mm
Length (Metric) 760 mm
Model 3850 ELP
Product Type Benchtop Autoclave
Temperature (English) 221°F to 278°F
Temperature (Metric) 105°C to 137°C
Weight (English) 220 lb.
Display Multi-color
Weight (Metric) 100 kg
Includes Stand, Trays, Printer, 3-Year Warranty
Printer Yes
Programmability Yes
Voltage 230 V
Sterility Non-sterile
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.