Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Tuttnauer™ Lab Line Benchtop Autoclave
SDP

Catalog No. 13890131 Shop All Heidolph North America Products
Change view
Click to view available options
Capacity (Metric):
28 L
65 L
85 L
160 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13-890-131 28 L
13-890-051 65 L
13-890-132 85 L
13-876-712 160 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 13-890-131 Supplier Heidolph™ Supplier No. 023210671
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Requires

Distilled Water

Benchtop autoclave designed to sterilize glassware, plastics, liquids and media.

Standard Operating Features

  • Designed to provide high quality, repeatable performance and accountability for a wide range of applications used in modern laboratories
  • Ideal for laboratory applications in research institutes, universities, pharmaceutical, biotechnology, food, and chemical industries
  • Benchtop lab autoclaves save space on crowded laboratory workbenches
  • For use with Agar preparation as well as the sterilization of liquids, pipettes, glass, instruments, and biohazards with various cooling options
  • Two integrated temperature probes for complete sterilization of liquids
  • Four preprogrammed cycles – glass, plastic, liquid A, and liquid B
  • Create up to 16 custom cycles and store up to 200 past cycles that you can open on your PC
  • Temperature selection from 105°C - 137°C (221°F - 279°F)
  • Water Saving System recycles used water after every cycle
  • Plug-n-play design: just add water to the reservoir and plug it in
  • Does not require connection to building utilities
  • PID (Proportional Integral Differential) pressure control
  • Built-in memory stores the previous 200 cycles
  • Multi-color display for easier reading, indicates stage of the cycle
  • Quick access to important information
  • Purchase includes autoclave stand, trays and printer

Service Plan

Extended warranty available

Warranty and Services

3-year warranty

Specifications

Certifications/Compliance UL, cUL, ASME
Capacity (English) 7.40 gal.
Capacity (Metric) 28 L
Depth (Metric) Chamber 46 cm
Diameter (English) Chamber 11 in.
Depth (English) Chamber 18 in.
Diameter (Metric) Chamber 28 cm
For Use With (Application) Sterilization of liquids, media, instruments, glassware
For Use With (Equipment) Laboratory Glassware and accessories
Length (English) 39.37 in.
Height (English) Exterior 31.50 in.
Height (Metric) Exterior 80 cm
Length (Metric) 100 cm
Model 2840ELP
Product Type Sterilizer
Temperature (English) 203°F to 219.2°F
Temperature (Metric) 95°C to 104°C
Weight (English) 112.42 lb.
Weight (Metric) 51 kg
Includes Stand, Trays, Printer, 3-Year Warranty
Printer Yes
Voltage 230 V
Sterility Non-sterile
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.