Learn More
Waters Corp TUV Detector Performance Maintenance Kit
Catalog No. 50818996 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit for the 2487 Dual Wavelength Absorbance Detector/2488 Multichannel Absorbance Detector/ACQUITY TUV Detector (through serial number K05UPT699N) contains all the Waters Quality Parts necessary to keep your detector operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts: a PerformancePLUS Long Life Lamp which is warranteed for 2000 hours or 1 year, whichever comes first.
Catalog No. 50-818-996 Supplier Waters Corp Supplier No. WAS081142
The Waters Performance Maintenance Kit for the 2487 Dual Wavelength Absorbance Detector/2488 Multichannel Absorbance Detector/ACQUITY TUV Detector (through serial number K05UPT699N) contains all the Waters Quality Parts necessary to keep your detector operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts: a PerformancePLUS Long Life Lamp which is warranteed for 2000 hours or 1 year, whichever comes first.