Learn More
MSC Tiger-Vac NFPA-652 Continuous Duty HEPA Vacuum, 26 gal, 1 ea
Catalog No. 502868997
Encompass
The Tiger-Vac NFPA-652 vacuum cleaner is a continuous duty, electric-powered unit designed for applications requiring HEPA filtration. It features a 26 gal stainless steel tank and is driven by a 3.2 hp motor.
  • NFPA-652 compliant and continuous duty operation
  • Electric power source with 3.2 hp motor and 15.8A draw
  • HEPA filtration for fine particulate capture
  • Maximum air flow of 270 CFM
  • Durable stainless steel tank with 26 gal capacity
  • Canister collection with disposable poly liner
  • Sound level of 70.0 dB for quiet operation
  • Includes comprehensive SD tool kit with 15 ft hose
Catalog No. 50-286-8997 Supplier MSC Supplier No. 29033883
Add to Cart
Add to Cart
Request a Quote
The Tiger-Vac NFPA-652 vacuum cleaner is a continuous duty, electric-powered unit designed for applications requiring HEPA filtration. It features a 26 gal stainless steel tank and is driven by a 3.2 hp motor.
  • NFPA-652 compliant and continuous duty operation
  • Electric power source with 3.2 hp motor and 15.8A draw
  • HEPA filtration for fine particulate capture
  • Maximum air flow of 270 CFM
  • Durable stainless steel tank with 26 gal capacity
  • Canister collection with disposable poly liner
  • Sound level of 70.0 dB for quiet operation
  • Includes comprehensive SD tool kit with 15 ft hose