Learn More
Selleck Chemical LLC TVB-3664 5mg
SDP
Supplier:  Selleck Chemical LLC S85635MG
Encompass_Preferred
TVB-3664 is a potent, selective, orally active and reversible inhibitor of fatty acid synthase (FASN) with IC50 of 0.018 ?M and 0.012 ?M for human and mouse cell palmitate synthesis, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3372