Learn More
Apexbio Technology LLC TW-37, 10mg. Cas: 877877-35-5 MFCD: MFCD17010275. Bcl-2 inhibitor, inhibits Bcl-2,Bcl-XL and Mcl-1
SDP
Catalog No. 501904910
Encompass_Preferred
TW-37 is a potent small-molecule inhibitor of BCL-2 (Ki = 290 nmol/L), which attenuates BCL-2 activation and inhibits multiple BCL-2 family members including BCL-XL (Ki = 1,110 nmol/L) and MCL-1 (Ki = 260 nmol/L). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4910 Supplier Apexbio Technology LLC Supplier No. A423410
TW-37 is a potent small-molecule inhibitor of BCL-2 (Ki = 290 nmol/L), which attenuates BCL-2 activation and inhibits multiple BCL-2 family members including BCL-XL (Ki = 1,110 nmol/L) and MCL-1 (Ki = 260 nmol/L). Other sizes are available. Please inqury us for quote.