Learn More
Abcam TW-37, 10MG
Catalog No. 502682952
Encompass_Preferred
MW 573.7 Da, Purity >98%. Small-molecule inhibitor of BCL-2 (Ki = 0.29 μM). Inhibits multiple BCL-2 family members including BCL-XL (Ki = 1.11 μM) and MCL-1 (Ki= 0.26 μM). Binds to the BCL-2 homology domain 3 (BH3) groove of BCL-2. Significant anti-proliferative and pro-apoptotic activity in lymphoma cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2952 Supplier Abcam Supplier No. AB27349010MG
MW 573.7 Da, Purity >98%. Small-molecule inhibitor of BCL-2 (Ki = 0.29 μM). Inhibits multiple BCL-2 family members including BCL-XL (Ki = 1.11 μM) and MCL-1 (Ki= 0.26 μM). Binds to the BCL-2 homology domain 3 (BH3) groove of BCL-2. Significant anti-proliferative and pro-apoptotic activity in lymphoma cells.
The product is subject to the following: Abcam Restricted Use Statement