Learn More
Medchemexpress LLC HY-12020 50mg Medchemexpress, TW-37 CAS:877877-35-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1202050MG
Encompass_Preferred
Medchemexpress, HY-12020 50mg TW-37 CAS:877877-35-5 TW-37 is a potent Bcl-2 inhibitor with Ki values of 260, 290 and 1110 nM for Mcl-1, Bcl-2 and Bcl-xL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9593