Learn More
Selleck Chemical LLC TW-37 10mg 877877-35-5
SDP
Catalog No. 508739
Encompass_Preferred
TW-37 is a novel nonpeptide inhibitor to recombinant Bcl-2, Bcl-xL and Mcl-1 with Ki of 0.29 ?M, 1.11 ?M and 0.26 ?M in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-873-9 Supplier Selleck Chemical LLC Supplier No. S112110MG
TW-37 is a novel nonpeptide inhibitor to recombinant Bcl-2, Bcl-xL and Mcl-1 with Ki of 0.29 ?M, 1.11 ?M and 0.26 ?M in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.