Learn More
Medchemexpress LLC HY-B0744B 100mg Medchemexpress, Eflornithine (hydrochloride, hydrate) CAS:96020-91-6 Purity:>98%
SDP
Catalog No. 501970123
Encompass_Preferred
Medchemexpress, HY-P7309 10ug TWEAK (CHO-expressed), Human CAS: Recombinant Human TWEAK (CHO-expressed) could induce apoptosis via multiple pathways of cell death in a cell type-specific manner and promote proliferation and migration of endothelial cells, and thus acts as a regulator of angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0123 Supplier Medchemexpress LLC Supplier No. HYB0744B100MG
Medchemexpress, HY-P7309 10ug TWEAK (CHO-expressed), Human CAS: Recombinant Human TWEAK (CHO-expressed) could induce apoptosis via multiple pathways of cell death in a cell type-specific manner and promote proliferation and migration of endothelial cells, and thus acts as a regulator of angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.