Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse TWEAK (TNFSF12) ELISA Kit

Catalog No. 502741227
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-274-1227 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-274-1227 Supplier Invitrogen™ Supplier No. EMTNFSF12X10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

The Mouse TWEAK (TNFSF12) ELISA quantitates Ms TWEAK in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms TWEAK. Principle of the method The Mouse TWEAK solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TNF-related weak inducer of apoptosis (TWEAK) is a member of the tumor necrosis factor superfamily (TNFSF) of structurally related cytokines. Like most other members of this family, TWEAK is a cell surface-associated type II transmembrane protein although a smaller, biologically active form can also be generated by cleavage near the cell membrane. TWEAK has multiple biological activities, including stimulation of cell growth and angiogenesis, induction of inflammatory cytokines, in addition to stimulation of apoptosis. The TWEAK signal transduction pathway has not been well established but it appears to signal via TweakR/Fn14 in a manner similar to that described for other TNFSF members that bind receptors lacking death domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O54907
Assay Range 0.27-200 pg/mL
Assay Sensitivity 0.3 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias APO3 ligand; APO3/DR3 ligand; APO3L; Dr3l; Dr3lg; LOW QUALITY PROTEIN: tumor necrosis factor ligand superfamily member 12; MGC129581; MGC20669; TNF superfamily member 12; TNF-related weak inducer of apoptosis; TNFSF12; TNLG4A; tumor necrosis factor (ligand) superfamily member 12; tumor necrosis factor (ligand) superfamily, member 12; tumor necrosis factor ligand 4A; tumor necrosis factor ligand superfamily member 12; Tumor necrosis factor ligand superfamily member 12, membrane form; Tumor necrosis factor ligand superfamily member 12, secreted form; tumor necrosis factor superfamily member 12; TWEAK; UNQ181/PRO207; zgc:153941APO3 ligand; APO3/DR3 ligand; APO3L; Dr3l; Dr3lg; LOW QUALITY PROTEIN: tumor necrosis factor ligand superfamily member 12; MGC129581; MGC20669; TNF superfamily member 12; TNF-related weak inducer of apoptosis; TNFSF12; TNLG4A; tumor necrosis factor (ligand) superfamily member 12; tumor necrosis factor (ligand) superfamily, member 12; tumor necrosis factor ligand 4A; tumor necrosis factor ligand superfamily member 12; Tumor necrosis factor ligand superfamily member 12, membrane form; Tumor necrosis factor ligand superfamily member 12, secreted form; tumor necrosis factor superfamily member 12; TWEAK; UNQ181/PRO207; zgc:153941; Apo3l; Dr3l; Dr3lg; Tweak
Gene ID (Entrez) 21944
Gene Symbol Tnfsf12
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases TNF-related weak inducer of apoptosis, TWEAK
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target TWEAK
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.