Learn More
ACROBiosystems ACROBiosystems Human TWEAK R / TNFRSF12A Protein, Fc Tag
Catalog No. 501957026
Encompass_Preferred
Recombinant Protein;100UG;Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK293). It contains AA Glu 28 - Trp 79 (Accession # AAH02718).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 32.5 kDa. As a result of glycosylation, the protein migrates as 32-45 kDa under reducing (R) condition, and 55-80 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-195-7026 Supplier ACROBiosystems Supplier No. TNAH526A100UG
Add to Cart
Add to Cart
Recombinant Protein;100UG;Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK293). It contains AA Glu 28 - Trp 79 (Accession # AAH02718).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 32.5 kDa. As a result of glycosylation, the protein migrates as 32-45 kDa under reducing (R) condition, and 55-80 kDa under non-reducing (NR) condition (SDS-PAGE).