Learn More
ACROBiosystems Recombinant Protein;Human TWEAK R / TNFRSF12 Protein, Fc Tag;HEK293;1MG;TNA,TNFRSF12A,FGF-inducible 14,FN14,TweakR,CD266,TNFRSF-12A,FN-14
Catalog No. 501658359
Encompass_Preferred
Recombinant Protein;Human TWEAK R / TNFRSF12 Protein, Fc Tag;1MG;Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK293). It contains AA Glu 28 - Trp 79 (Accession # AAH02718).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 33 kDa. As a result of glycosylation, the protein migrates as 32-40 kDa under reducing (R) condition, and 65-80 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8359 Supplier ACROBiosystems Supplier No. TNAH526A1MG
Recombinant Protein;Human TWEAK R / TNFRSF12 Protein, Fc Tag;1MG;Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK293). It contains AA Glu 28 - Trp 79 (Accession # AAH02718).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 33 kDa. As a result of glycosylation, the protein migrates as 32-40 kDa under reducing (R) condition, and 65-80 kDa under non-reducing (NR) condition (SDS-PAGE).