Learn More
Sigma Aldrich Fine Chemicals Biosciences TWEEN 20 | 9005-64-5 | MFCD00165986 | 1 L
Catalog No. NC1451069
Encompass
TWEEN 20 | Purity: ≥40.0% | Mol Wt: ~1228 | 9005-64-5 | MFCD00165986 | 1 L
Catalog No. NC1451069 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P13791L
TWEEN 20 | Purity: ≥40.0% | Mol Wt: ~1228 | 9005-64-5 | MFCD00165986 | 1 L