Learn More
Selleck Chemical LLC Tween-20-100mL
SDP
Supplier:  Selleck Chemical LLC S4621
Encompass_Preferred
Tween-20 (Polysorbates 20) is used in the formulation of biotherapeutic products for both preventing surface adsorption and as a stabilizer against protein aggregation.
Catalog No. 50-238-9917