Learn More
Selleck Chemical LLC Tween-20 S4621-100mL
SDP
Catalog No. 503110806
Encompass_Preferred
Tween-20 (Polysorbates 20) is used in the formulation of biotherapeutic products for both preventing surface adsorption and as a stabilizer against protein aggregation
Catalog No. 50-311-0806 Supplier Selleck Chemical LLC Supplier No. S4621100ML
Tween-20 (Polysorbates 20) is used in the formulation of biotherapeutic products for both preventing surface adsorption and as a stabilizer against protein aggregation