Learn More
Research Products International Corp Tween 20 [Polyoxyethylenesorbitan Monolaurate], 500 Milliliters
SDP
Catalog No. 50213612
Encompass_Preferred
Fatty acid composition:
Lauric acid: Approximately 50%
Balance: Myristic, Palmitic and Stearic acids.
Catalog No. 50-213-612 Supplier Research Products International Corp Supplier No. P203700.5
Fatty acid composition:
Lauric acid: Approximately 50%
Balance: Myristic, Palmitic and Stearic acids.