Learn More
Biotium Tween 20
Catalog No. 501964389
Encompass_Preferred
Tween-20 (polysorbate 20) is a polysorbate surfactant commonly used as a blocking and washing agent in Western blotting and other immunostaining assays. Tween-20 is typically included in blocking buffers and washing buffers at a concentration of 0.1% to reduce non-specific antibody binding. TWEEN is a registered trademark of Uniqema Americas LLC.
Catalog No. 50-196-4389 Supplier Biotium Supplier No. 22002
Tween-20 (polysorbate 20) is a polysorbate surfactant commonly used as a blocking and washing agent in Western blotting and other immunostaining assays. Tween-20 is typically included in blocking buffers and washing buffers at a concentration of 0.1% to reduce non-specific antibody binding. TWEEN is a registered trademark of Uniqema Americas LLC.