Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tween™ 20, For Synthesis, MilliporeSigma™

Catalog No. M8221840050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
1 L
2.5 L
Packaging:
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8221840050 50 mL Plastic Bottle
M8221840500 500 mL Plastic Bottle
M8221841000 1 L Plastic Bottle
M8221842500 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M8221840050 Supplier MilliporeSigma Supplier No. 8.22184.0050
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
PubChem CID 443314
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Physical Form Liquid
Boiling Point 100°C
Density 1.1 g/3 (25 °C)
pH 6.0 to 8.0
Quantity 50 mL
Flash Point 275°C
Packaging Plastic Bottle
Solubility Information <0.2mg/L soluble
Formula Weight 1228
EINECSNumber 500-018-3
RTECSNumber TR7400000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.