Learn More
Enzo Life Sciences Tween-20 solution (10%), (30 ml) (30 ml)
SDP
Catalog No. 50658090
Encompass_Preferred
This Tween-20 solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in IMMUNOSET products. Each bottle is sufficient for use with 5x96 wells microtiter plates if following the instructions in our IMMUNOSET protocol. The Tween-20 solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our IMMUNOSET products. Shipping: Ambient. Long Term Storage: +4°C
Catalog No. 50-658-090 Supplier Enzo Life Sciences Supplier No. ADI801929
This Tween-20 solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in IMMUNOSET products. Each bottle is sufficient for use with 5x96 wells microtiter plates if following the instructions in our IMMUNOSET protocol. The Tween-20 solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our IMMUNOSET products. Shipping: Ambient. Long Term Storage: +4°C