Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tween 20™, Ultrapure, Thermo Scientific Chemicals

Catalog No. p-7082764
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ20605AP 500 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ20605AP Supplier Thermo Scientific Supplier No. J20605AP
Only null left
Add to Cart

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylene sorbitan monolaurate
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Physical Form Liquid
Boiling Point >100°C
Density 1.105
Quantity 500 mL
Flash Point >110°C
Packaging Plastic bottle
Specific Gravity 1.105
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Grade Ultra Pure
Chemical Name or Material Tween 20™
EINECSNumber 500-018-3
RTECSNumber TR7400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.