Learn More
Medchemexpress LLC HY-Y1891 100mL Medchemexpress, Tween 80 CAS:9005-65-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY1891100ML
Encompass_Preferred
Medchemexpress, HY-Y1891 100mL Tween 80 CAS:9005-65-6 Tween 80 is a surfactant which can also reduce bacterial attachment and inhibit biofilm formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9442