Learn More
Selleck Chemical LLC Tween 80 | 99% | 9005-65-6 | 100mL
SDP
Catalog No. NC2023234
Encompass_Preferred
Tween 80 is a polysorbate surfactant used for selective protein extraction and the isolation of nuclei from mammalian cell lines. Tween 80 has been used in drug vehicle solutions for specific injections in mice and rats. It has been used for the determination of esterase activity in bacterial cells..
Catalog No. NC2023234 Supplier Selleck Chemical LLC Supplier No. S6702100ML
Tween 80 is a polysorbate surfactant used for selective protein extraction and the isolation of nuclei from mammalian cell lines. Tween 80 has been used in drug vehicle solutions for specific injections in mice and rats. It has been used for the determination of esterase activity in bacterial cells..