Learn More
Research Products International Corp Tween 80 (Polyoxyethylenesorbitan, Monooleate), 4 Liters
SDP
Catalog No. 501368496
Encompass_Preferred
Fatty acid compostion: Oleic acid approximately 70%
Balance primarily linoleic; palmitic and stearic acids.
Catalog No. 50-136-8496 Supplier Research Products International Corp Supplier No. P203904.0
Fatty acid compostion: Oleic acid approximately 70%
Balance primarily linoleic; palmitic and stearic acids.